accessibility menu, dialog, popup

UserName

CAS RN 103879-58-9

CH3NOOOH

CAS RN 103879-58-9

IUPAC Name: 5-methyl-1,3-oxazole-4-carboxylic acid
Synonyms: 5-methyl-1,3-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 127.1
Molecular Formula: C5H5NO3
InChi Key: QIACATCUODRSLS-UHFFFAOYSA-N
SMILES: CC1=C(N=CO1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results