CAS RN 104098-48-8
CAS RN 104098-48-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Imazapic, MedChemExpress
MedChemExpress Imazapic is a selective herbicide for both the preand post-emergent control of some annual and perennial grasses and some broadleaf weeds

