CAS RN 1043-21-6
CAS RN 1043-21-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pirenoxine, TRC
CAS: 1043-21-6 Molecular Formula: C16H8N2O5 Molecular Weight (g/mol): 308.25 Synonym: 1-Hydroxy-3-carboxy-5H-pyrido[3,2-a]phenoxazin-5-one 1-Hydroxy-5H-pyrido[3,2-a]phenoxazin-5-one-3-carboxylic acid,Catalin,Catalin (pharmaceutical),Catalin K,Clarvisan,HPPC,Kary-Uni,Pirfenoxone,Pyrphenoxone IUPAC Name: 1-hydroxy-5-oxopyrido[3,2-a]phenoxazine-3-carboxylic acid SMILES: O=C1C2=NC(C(O)=O)=CC(O)=C2C3=NC4=C(C=CC=C4)OC3=C1