accessibility menu, dialog, popup

UserName

CAS RN 104376-24-1

BH3CHOOH(E/Z)

CAS RN 104376-24-1

IUPAC Name: (pent-1-en-1-yl)boronic acid
Synonyms: pent-1-en-1-ylboronic acid
Molecular Weight (g/mol): 113.95
Molecular Formula: C5H11BO2
InChi Key: SYKWJOZHNDPWIM-UHFFFAOYSA-N
SMILES: CCCC=CB(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results