accessibility menu, dialog, popup

Test

CAS RN 1048973-47-2

CAS RN 1048973-47-2

IUPAC Name: N'-(1-hexyl-2-oxo-2,3-dihydro-1H-indol-3-ylidene)benzohydrazide
Synonyms: N'-(1-hexyl-2-oxoindol-3-ylidene)benzohydrazide
Molecular Weight (g/mol): 349.43
Molecular Formula: C21H23N3O2
InChi Key: ZGQHMZCITJHYOW-UHFFFAOYSA-N
SMILES: CCCCCCN1C(=O)C(=NNC(=O)C2=CC=CC=C2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results