CAS RN 10490-07-0
IUPAC Name:
2-phenyl-2-(phenylsulfanyl)acetic acid
Synonyms:
phenyl(phenylsulfanyl)acetic acid
Molecular Weight (g/mol):
244.31
Molecular Formula:
C14H12O2S
InChi Key:
UOCQATUFLPHRNG-UHFFFAOYNA-N
SMILES:
OC(=O)C(SC1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
alpha-(Phenylthio)phenylacetic acid, 99%, Thermo Scientific™
CAS: 10490-07-0 Molecular Formula: C14H12O2S Molecular Weight (g/mol): 244.308 MDL Number: MFCD00066259 InChI Key: UOCQATUFLPHRNG-UHFFFAOYSA-N Synonym: phenyl phenylsulfanyl acetic acid,2-phenyl-2-phenylthio acetic acid,alpha-phenylthio phenylacetic acid,2-phenyl-2-phenylsulfanyl acetic acid,2-phenyl-2-phenylthioacetic acid,acmc-20alu8,phenylthio phenylacetic acid,phenylthio-phenyl acetic acid,phenyl phenylthio acetic acid,alpha-phenylthio-phenylacetic acid PubChem CID: 239605 IUPAC Name: 2-phenyl-2-phenylsulfanylacetic acid SMILES: C1=CC=C(C=C1)C(C(=O)O)SC2=CC=CC=C2