CAS RN 105088-08-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dexamethasone Beta-D-Glucuronide Sodium Salt, TRC
CAS: 105088-08-2 Molecular Formula: C28 H36 F Na O11 Molecular Weight (g/mol): 590.57 SMILES: [Na+].C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)[O-]