CAS RN 1055301-16-0
IUPAC Name:
1,2-bis(2-chlorophenyl)ethane-1,2-diamine dihydrochloride
Synonyms:
1,2-bis(2-chlorophenyl)ethane-1,2-diamine dihydrochloride
Molecular Weight (g/mol):
354.1
Molecular Formula:
C14H16Cl4N2
InChi Key:
YXGVUHANCVCDJJ-UHFFFAOYNA-N
SMILES:
Cl.Cl.NC(C(N)C1=CC=CC=C1Cl)C1=CC=CC=C1Cl
1
–
1
of
1
results
1
(R,R)-1,2-Bis(2-chlorophenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%, Thermo Scientific™
CAS: 1055301-16-0 Molecular Formula: C14H16Cl4N2 Molecular Weight (g/mol): 354.10 MDL Number: MFCD09265294,MFCD09953699 InChI Key: YXGVUHANCVCDJJ-UHFFFAOYNA-N Synonym: r,r-bis-2-chlorophenyl ethylenediamine dihydrochloride,r,r-1,2-bis 2-chlorophenyl-1,2-ethanediamine dihydrochloride,1r,2r-1,2-bis 2-chlorophenyl ethane-1,2-diamine dihydrochloride,r,r-1,2-bis 2-chlorophenyl-1,2-ethanediamine dihydrochloride, ee PubChem CID: 73995086 IUPAC Name: (1R,2R)-1,2-bis(2-chlorophenyl)ethane-1,2-diamine;dihydrochloride SMILES: Cl.Cl.NC(C(N)C1=CC=CC=C1Cl)C1=CC=CC=C1Cl