CAS RN 1055301-17-1
IUPAC Name:
1,2-bis(naphthalen-1-yl)ethane-1,2-diamine dihydrochloride
Synonyms:
1,2-bis(naphthalen-1-yl)ethane-1,2-diamine dihydrochloride
Molecular Weight (g/mol):
385.33
Molecular Formula:
C22H22Cl2N2
InChi Key:
SHNGCXWOHADIKG-UHFFFAOYNA-N
SMILES:
Cl.Cl.NC(C(N)C1=C2C=CC=CC2=CC=C1)C1=CC=CC2=CC=CC=C12
1
–
1
of
1
results
1
(R,R)-1,2-Di(1-naphthyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%, Thermo Scientific™
CAS: 1055301-17-1 Molecular Formula: C22H22Cl2N2 Molecular Weight (g/mol): 385.332 MDL Number: MFCD09265310 InChI Key: SHNGCXWOHADIKG-XZHZQXPNSA-N Synonym: r,r-bis-1-naphthyl ethylenediamine dihydrochloride,r,r-bis-4-methoxyphenyl ethylenediamine dihydrochloride,r,r-1,2-bis 1-naphthyl-1,2-ethanediamine dihydrochloride,r,r-1,2-di 1-naphthyl-1,2-ethanediamine dihydrochloride,1r,2r-1,2-bis naphthalen-1-yl ethane-1,2-diamine dihydrochloride,r,r-1,2-di 1-naphthyl-1,2-ethanediamine dihydrochloride, ee PubChem CID: 45925673 IUPAC Name: (1R,2R)-1,2-dinaphthalen-1-ylethane-1,2-diamine;dihydrochloride SMILES: C1=CC=C2C(=C1)C=CC=C2C(C(C3=CC=CC4=CC=CC=C43)N)N.Cl.Cl