accessibility menu, dialog, popup

UserName

CAS RN 105614-25-3

FFH2NHCl

CAS RN 105614-25-3

IUPAC Name: 2,2-difluorocyclopropan-1-amine hydrochloride
Synonyms: 2,2-difluorocyclopropan-1-amine hydrochloride
Molecular Weight (g/mol): 129.53
Molecular Formula: C3H6ClF2N
InChi Key: WSSNWLHFBOKGGL-UHFFFAOYNA-N
SMILES: Cl.NC1CC1(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)