CAS RN 10606-46-9
CAS RN 10606-46-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
o,p’-Dicofol, TRC
CAS: 10606-46-9 Molecular Formula: C14 H9 Cl5 O Molecular Weight (g/mol): 370.49 Synonym: Benzhydrol, 2,4'-dichloro-α-(trichloromethyl)- (7CI),2-Chloro-α-(4-chlorophenyl)-α-(trichloromethyl)benzenemethanol,o,p'-Dicofol IUPAC Name: 2,2,2-trichloro-1-(2-chlorophenyl)-1-(4-chlorophenyl)ethanol SMILES: OC(c1ccc(Cl)cc1)(c2ccccc2Cl)C(Cl)(Cl)Cl