accessibility menu, dialog, popup

UserName

CAS RN 107133-36-8

CH3(S)NH(S)CH3ON(S)(S)(S)HOOOOCH3CH3CH3CH3H2N

CAS RN 107133-36-8

IUPAC Name: (2S,3aS,7aS)-1-[(2S)-2-{[(2S)-1-ethoxy-1-oxopentan-2-yl]amino}propanoyl]-octahydro-1H-indole-2-carboxylic acid; 2-methylpropan-2-amine
Synonyms: aceon
Molecular Weight (g/mol): 441.61
Molecular Formula: C23H43N3O5
InChi Key: IYNMDWMQHSMDDE-MHXJNQAMSA-N
SMILES: CC(C)(C)N.CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(O)=O)C(=O)OCC
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results