accessibility menu, dialog, popup

UserName

CAS RN 1073354-26-3

BOOH3CCH3H3CH3CNNNOOCH3

CAS RN 1073354-26-3

IUPAC Name: ethyl 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine-1-carboxylate
Synonyms: ethyl 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine-1-carboxylate
Molecular Weight (g/mol): 361.25
Molecular Formula: C18H28BN3O4
InChi Key: XKCNZNSTOYPVDQ-UHFFFAOYSA-N
SMILES: CCOC(=O)N1CCN(CC1)C1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results