accessibility menu, dialog, popup

UserName

CAS RN 1073372-07-2

BOOFF

CAS RN 1073372-07-2

IUPAC Name: 2-(2,4-difluorophenyl)-1,3,2-dioxaborinane
Synonyms: 2-(2,4-difluorophenyl)-1,3,2-dioxaborinane
Molecular Weight (g/mol): 197.98
Molecular Formula: C9H9BF2O2
InChi Key: AHKNGRYTXDKKDQ-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C=C1)B1OCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results