CAS RN 1083181-12-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 1-(Phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine-5-carboxylate, TRC
CAS: 1083181-12-7 Molecular Formula: C15H12N2O4S Molecular Weight (g/mol): 316.33 IUPAC Name: methyl 1-(benzenesulfonyl)pyrrolo[2,3-b]pyridine-5-carboxylate SMILES: COC(=O)c1cnc2c(ccn2S(=O)(=O)c3ccccc3)c1