CAS RN 108612-45-9
CAS RN 108612-45-9
IUPAC Name:
2-[(1-{1-[(4-fluorophenyl)methyl]-1H-1,3-benzodiazol-2-yl}piperidin-4-yl)(methyl)amino]-3,4-dihydropyrimidin-4-one
Synonyms:
mizolastine
Molecular Weight (g/mol):
432.5
Molecular Formula:
C24H25FN6O
InChi Key:
PVLJETXTTWAYEW-UHFFFAOYSA-N
SMILES:
CN(C1CCN(CC1)C1=NC2=CC=CC=C2N1CC1=CC=C(F)C=C1)C1=NC=CC(=O)N1
1
–
3
of
3
results
1
Mizolastine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Mizolastine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Mizolastine, MedChemExpress
MedChemExpress Mizolastine is a histamine H1-receptor antagonist with IC50 of 47 nM used in the treatment of hay fever (seasonal allergic rhinitis), hives and other allergic reactions.