accessibility menu, dialog, popup

UserName

CAS RN 1092460-83-7

FOOHOFFF

CAS RN 1092460-83-7

IUPAC Name: 5-fluoro-2-(trifluoromethoxy)benzoic acid
Synonyms: 5-fluoro-2-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 224.11
Molecular Formula: C8H4F4O3
InChi Key: JRQMLAPRSGFPRF-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(OC(F)(F)F)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results