accessibility menu, dialog, popup

UserName

CAS RN 110-01-0

S

CAS RN 110-01-0

IUPAC Name: thiolane
Synonyms: tetrahydrothiophene pennodorant 1073 pennodorant 1013 tetrahydrothiofen THT thilane thiofan thiophane
Molecular Weight (g/mol): 88.17
Molecular Formula: C4H8S
InChi Key: RAOIDOHSFRTOEL-UHFFFAOYSA-N
SMILES: C1CCSC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results