accessibility menu, dialog, popup

UserName

H3CSH

CAS RN 110-66-7

IUPAC Name: pentane-1-thiol
Synonyms: 1-pentanethiol
Molecular Weight (g/mol): 104.21
Molecular Formula: C5H12S
InChi Key: ZRKMQKLGEQPLNS-UHFFFAOYSA-N
SMILES: CCCCCS
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)