accessibility menu, dialog, popup

UserName

CAS RN 110553-27-0

H3CSCH3OHSCH3

CAS RN 110553-27-0

IUPAC Name: 2-methyl-4,6-bis[(octylsulfanyl)methyl]phenol
Synonyms: 2-methyl-4,6-bis[(octylsulfanyl)methyl]phenol
Molecular Weight (g/mol): 424.75
Molecular Formula: C25H44OS2
InChi Key: GAODDBNJCKQQDY-UHFFFAOYSA-N
SMILES: CCCCCCCCSCC1=CC(C)=C(O)C(CSCCCCCCCC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results