accessibility menu, dialog, popup

UserName

CH3CH3OHOOCH3H3CHO

CAS RN 1115-20-4

IUPAC Name: 3-hydroxy-2,2-dimethylpropyl 3-hydroxy-2,2-dimethylpropanoate
Synonyms: 3-hydroxy-2,2-dimethylpropyl 3-hydroxy-2,2-dimethylpropanoate
Molecular Weight (g/mol): 204.27
Molecular Formula: C10H20O4
InChi Key: SZCWBURCISJFEZ-UHFFFAOYSA-N
SMILES: CC(C)(CO)COC(=O)C(C)(C)CO
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results