accessibility menu, dialog, popup

UserName

CAS RN 111721-75-6

FBrH2N

CAS RN 111721-75-6

IUPAC Name: 2-bromo-3-fluoroaniline
Synonyms: 2-bromo-3-fluoroaniline
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: XZRSXRUYZXBTGD-UHFFFAOYSA-N
SMILES: NC1=C(Br)C(F)=CC=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results