accessibility menu, dialog, popup

UserName

H3CCH2

CAS RN 1120-36-1

IUPAC Name: tetradec-1-ene
Synonyms: tetradecene
Molecular Weight (g/mol): 196.38
Molecular Formula: C14H28
InChi Key: HFDVRLIODXPAHB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC=C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results