accessibility menu, dialog, popup

UserName

CAS RN 1122-17-4

IUPAC Name: dichloro-2,5-dihydrofuran-2,5-dione
Synonyms: dichlorofuran-2,5-dione
Molecular Weight (g/mol): 166.94
Molecular Formula: C4Cl2O3
InChi Key: AGULWIQIYWWFBJ-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(=O)OC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results