accessibility menu, dialog, popup

UserName

CAS RN 112281-77-3

FFFFONNNClCl

CAS RN 112281-77-3

IUPAC Name: 1-[2-(2,4-dichlorophenyl)-3-(1,1,2,2-tetrafluoroethoxy)propyl]-1H-1,2,4-triazole
Synonyms: tetraconazole
Molecular Weight (g/mol): 372.14
Molecular Formula: C13H11Cl2F4N3O
InChi Key: LQDARGUHUSPFNL-UHFFFAOYNA-N
SMILES: FC(F)C(F)(F)OCC(CN1C=NC=N1)C1=CC=C(Cl)C=C1Cl
Concentration 
  • (3)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)