accessibility menu, dialog, popup

UserName

CAS RN 1123-25-7

H3COO

CAS RN 1123-25-7

IUPAC Name: 1-methylcyclohexane-1-carboxylate
Synonyms: 1-methylcyclohexane-1-carboxylate
Molecular Weight (g/mol): 141.19
Molecular Formula: C8H13O2
InChi Key: REHQLKUNRPCYEW-UHFFFAOYSA-M
SMILES: CC1(CCCCC1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results