accessibility menu, dialog, popup

UserName

CAS RN 112641-64-2

H3COClOF

CAS RN 112641-64-2

IUPAC Name: 6-chloro-2-fluoro-3-methoxybenzaldehyde
Synonyms: 6-chloro-2-fluoro-3-methoxybenzaldehyde
Molecular Weight (g/mol): 188.58
Molecular Formula: C8H6ClFO2
InChi Key: OADUJQWBRPRPQX-UHFFFAOYSA-N
SMILES: COC1=C(F)C(C=O)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results