CAS RN 1128165-88-7
CAS RN 1128165-88-7
IUPAC Name:
2-cyano-3-{1-[3-(dimethylamino)propyl]-1H-indol-3-yl}-N-octylprop-2-enamide
Synonyms:
2-cyano-3-{1-[3-(dimethylamino)propyl]indol-3-yl}-N-octylprop-2-enamide
Molecular Weight (g/mol):
408.59
Molecular Formula:
C25H36N4O
InChi Key:
MYIMRONQBLZJFI-UHFFFAOYSA-N
SMILES:
CCCCCCCCNC(=O)C(=CC1=CN(CCCN(C)C)C2=CC=CC=C12)C#N