accessibility menu, dialog, popup

UserName

CAS RN 1131-17-5

CH3ClNN

CAS RN 1131-17-5

IUPAC Name: 5-chloro-3-methyl-1-phenyl-1H-pyrazole
Synonyms: 5-chloro-3-methyl-1-phenylpyrazole
Molecular Weight (g/mol): 192.65
Molecular Formula: C10H9ClN2
InChi Key: ZZOWFLAMMWOSCG-UHFFFAOYSA-N
SMILES: CC1=NN(C(Cl)=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)