accessibility menu, dialog, popup

UserName

CAS RN 113170-72-2

NH2BrH2NFFF

CAS RN 113170-72-2

IUPAC Name: 3-bromo-5-(trifluoromethyl)benzene-1,2-diamine
Synonyms: 3-bromo-5-(trifluoromethyl)benzene-1,2-diamine
Molecular Weight (g/mol): 255.04
Molecular Formula: C7H6BrF3N2
InChi Key: OVGAPUVMLZWQQF-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(Br)=C1N)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)