accessibility menu, dialog, popup

UserName

CH3NH2OHO

CAS RN 1132-26-9

IUPAC Name: (2S,3S)-2-azaniumyl-3-phenylbutanoate
Synonyms: (2S,3S)-2-ammonio-3-phenylbutanoate
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: IRZQDMYEJPNDEN-CBAPKCEASA-N
SMILES: C[C@H]([C@H]([NH3+])C([O-])=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results