accessibility menu, dialog, popup

UserName

CAS RN 113451-59-5

H3CH3CCH3OONNH

CAS RN 113451-59-5

IUPAC Name: tert-butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
Synonyms: tert-butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
Molecular Weight (g/mol): 198.27
Molecular Formula: C10H18N2O2
InChi Key: UXAWXZDXVOYLII-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC2CC1CN2
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)