CAS RN 114213-69-3
CAS RN 114213-69-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Methyl Gatifloxacin, TRC
CAS: 114213-69-3 Molecular Formula: C20H24FN3O4 Molecular Weight (g/mol): 389.42 Synonym: 1-Cyclopropyl-7-(3,4-dimethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid IUPAC Name: 1-cyclopropyl-7-(3,4-dimethylpiperazin-1-yl)-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid SMILES: COc1c(N2CCN(C)C(C)C2)c(F)cc3C(=O)C(=CN(C4CC4)c13)C(=O)O