accessibility menu, dialog, popup

UserName

NHN

CAS RN 1145-01-3

IUPAC Name: 3,5-diphenyl-1H-pyrazole
Synonyms: 3,5-diphenyl-1H-pyrazole
Molecular Weight (g/mol): 220.28
Molecular Formula: C15H12N2
InChi Key: JXHKUYQCEJILEI-UHFFFAOYSA-N
SMILES: N1N=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)