accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CBrF

CAS RN 1150271-74-1

IUPAC Name: 2-[4-(bromomethyl)-3-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[4-(bromomethyl)-3-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 314.99
Molecular Formula: C13H17BBrFO2
InChi Key: OOTPRFIHAFABSN-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(F)=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results