accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNNOCH3

CAS RN 1150561-76-4

IUPAC Name: 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]ethan-1-one
Synonyms: 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]ethanone
Molecular Weight (g/mol): 236.08
Molecular Formula: C11H17BN2O3
InChi Key: GAXOQGDPSCOOLC-UHFFFAOYSA-N
SMILES: CC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)