CAS RN 1152091-69-4
CAS RN 1152091-69-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methyllamotrigine Methanesulfonate, TRC
CAS: 1152091-69-4 Molecular Formula: C10 H9 Cl2 N5 . C H4 O3 S Molecular Weight (g/mol): 366.22 Synonym: 3,5-Diamino-6-(2,3-dichlorophenyl)-2-methyl-1,2,4-triazinium Methane Sulfonate IUPAC Name: 6-(2,3-dichlorophenyl)-3-imino-2-methyl-1,2,4-triazin-5-amine;methanesulfonic acid SMILES: CN1N=C(C(=NC1=N)N)c2cccc(Cl)c2Cl.CS(=O)(=O)O