CAS RN 115534-33-3
CAS RN 115534-33-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress TMA-DPH
MedChemExpress TMA-DPH is a hydrophobic fluorescent membrane probe (Ex=355 nm; Em=430 nm).