CAS RN 1159-54-2
IUPAC Name:
tris(4-chlorophenyl)phosphane
Synonyms:
tris(4-chlorophenyl)phosphane
Molecular Weight (g/mol):
365.62
Molecular Formula:
C18H12Cl3P
InChi Key:
IQKSLJOIKWOGIZ-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)P(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
Tris(4-chlorophenyl)phosphine, 97%
CAS: 1159-54-2 Molecular Formula: C18H12Cl3P Molecular Weight (g/mol): 365.618 MDL Number: MFCD00013639 InChI Key: IQKSLJOIKWOGIZ-UHFFFAOYSA-N Synonym: tris 4-chlorophenyl phosphine,tri p-chlorophenyl phosphine,tris p-chlorophenyl phosphine,tris 4-chlorophenyl phosphane,tri-4-chlorophenyl phosphine,phosphine, tris 4-chlorophenyl,tri 4-chlorophenyl phosphine,acmc-20ajbt,pubchem6454 PubChem CID: 70874 IUPAC Name: tris(4-chlorophenyl)phosphane SMILES: C1=CC(=CC=C1P(C2=CC=C(C=C2)Cl)C3=CC=C(C=C3)Cl)Cl