accessibility menu, dialog, popup

UserName

CAS RN 1164500-72-4

CH3ONSSOHNOOHOH(E/Z)(E/Z)

CAS RN 1164500-72-4

IUPAC Name: 2-hydroxy-4-{4-[5-(2-methyl-3-phenylprop-2-en-1-ylidene)-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]butanamido}benzoic acid
Synonyms: 2-hydroxy-4-{4-[5-(2-methyl-3-phenylprop-2-en-1-ylidene)-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]butanamido}benzoic acid
Molecular Weight (g/mol): 482.57
Molecular Formula: C24H22N2O5S2
InChi Key: COFMYJWNXSFLKQ-UHFFFAOYSA-N
SMILES: CC(C=C1SC(=S)N(CCCC(=O)NC2=CC(O)=C(C=C2)C(O)=O)C1=O)=CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results