accessibility menu, dialog, popup

UserName

CAS RN 116628-53-6

BOOCH3CH3NH2

CAS RN 116628-53-6

IUPAC Name: diphenylboranyl 2-amino-4-methylpentanoate
Synonyms: diphenylboranyl 2-amino-4-methylpentanoate
Molecular Weight (g/mol): 295.19
Molecular Formula: C18H22BNO2
InChi Key: HEFYWEFHUCZFOZ-UHFFFAOYNA-N
SMILES: CC(C)CC(N)C(=O)OB(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results