CAS RN 117186-02-4
IUPAC Name:
3-methyl-5-(trifluoromethyl)benzoic acid
Synonyms:
3-methyl-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol):
204.15
Molecular Formula:
C9H7F3O2
InChi Key:
QJSRENHDYMAJIL-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC(=C1)C(O)=O)C(F)(F)F
1
–
1
of
1
results
1
3-Methyl-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™
CAS: 117186-02-4 Molecular Formula: C9H7F3O2 Molecular Weight (g/mol): 204.148 MDL Number: MFCD01631590 InChI Key: QJSRENHDYMAJIL-UHFFFAOYSA-N Synonym: 3-methyl-5-trifluoromethyl benzoic acid,3-methyl-5-trifluoromethyl-benzoic acid,benzoic acid, 3-methyl-5-trifluoromethyl,3-carboxy-5-methylbenzotrifluoride,3-methyl-5-trifluoromethyl benzoicacid,pubchem16302,acmc-1bpp2,ksc495m6n,5-methyl-3-trifluoromethyl benzoic acid PubChem CID: 2775588 IUPAC Name: 3-methyl-5-(trifluoromethyl)benzoic acid SMILES: CC1=CC(=CC(=C1)C(=O)O)C(F)(F)F