CAS RN 118443-89-3
CAS RN 118443-89-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Cefquinome Sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Cefquinome Sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Cefquinome Sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.