CAS RN 1186195-62-9
CAS RN 1186195-62-9
IUPAC Name:
4-(1-benzothiophen-2-yl)-N-{4-[4-(pyrrolidin-1-yl)piperidine-1-carbonyl]phenyl}pyrimidin-2-amine hydrochloride
Synonyms:
4-(1-benzothiophen-2-yl)-N-{4-[4-(pyrrolidin-1-yl)piperidine-1-carbonyl]phenyl}pyrimidin-2-amine hydrochloride
Molecular Weight (g/mol):
520.09
Molecular Formula:
C28H30ClN5OS
InChi Key:
XFVDIVQCZWDBCW-UHFFFAOYSA-N
SMILES:
Cl.O=C(N1CCC(CC1)N1CCCC1)C1=CC=C(NC2=NC=CC(=N2)C2=CC3=CC=CC=C3S2)C=C1