CAS RN 1187-84-4
IUPAC Name:
2-amino-3-(methylsulfanyl)propanoic acid
Synonyms:
methylcysteine acm-thiopropionate S-methyl-(9CI)-L-cysteine L-methylcysteine acm-thiopropionic acid
Molecular Weight (g/mol):
135.18
Molecular Formula:
C4H9NO2S
InChi Key:
IDIDJDIHTAOVLG-UHFFFAOYNA-N
SMILES:
CSCC(N)C(O)=O
1
–
2
of
2
results
1
S-Methyl-L-cysteine, TRC
CAS: 1187-84-4 Molecular Formula: C4 H9 N O2 S Molecular Weight (g/mol): 135.19 Synonym: S-Methylcysteine,L-Methylcysteine,H-Cys(Me)-OH,(R)-2-amino-3-(methylthio)propanoic acid,3-(Methylthio)-L-alanine,(R)-2-Amino-3-(methylmercapto)propionic acid,(-)-S-Methyl-L-cysteine IUPAC Name: (2R)-2-amino-3-methylsulfanyl-propanoic acid SMILES: CSC[C@H](N)C(=O)O
2
S-Methyl-L-cysteine, MedChemExpress
MedChemExpress S-Methyl-L-cysteine is a natural product that acts as a substrate in the catalytic antioxidant system mediated by methionine sulfoxide reductase A (MSRA), with antioxidative, neuroprotective, and anti-obesity activities.