accessibility menu, dialog, popup

UserName

CAS RN 1191-25-9

IUPAC Name: 6-hydroxyhexanoate
Synonyms: 6-hydroxyhexanoate
Molecular Weight (g/mol): 131.15
Molecular Formula: C6H11O3
InChi Key: IWHLYPDWHHPVAA-UHFFFAOYSA-M
SMILES: OCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results