accessibility menu, dialog, popup

UserName

NNOH(E/Z)

CAS RN 1193-96-0

IUPAC Name: 2-(nitrosomethylidene)-1,2-dihydropyridine
Synonyms: 2-(nitrosomethylidene)-1H-pyridine
Molecular Weight (g/mol): 122.13
Molecular Formula: C6H6N2O
InChi Key: VRGKDCRTYBCJTA-UHFFFAOYSA-N
SMILES: O=NC=C1NC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)