accessibility menu, dialog, popup

UserName

CAS RN 1198097-97-0

ONSH2NHO(E/Z)

CAS RN 1198097-97-0

IUPAC Name: 2-amino-5-[(4-hydroxyphenyl)methylidene]-4,5-dihydro-1,3-thiazol-4-one
Synonyms: 2-amino-5-[(4-hydroxyphenyl)methylidene]-1,3-thiazol-4-one
Molecular Weight (g/mol): 220.25
Molecular Formula: C10H8N2O2S
InChi Key: YBHQCJILTOVLHD-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C(S1)=CC1=CC=C(O)C=C1
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)