CAS RN 120-72-9
CAS RN 120-72-9
IUPAC Name:
1H-indole
Synonyms:
indole 2,3-benzopyrrole 14c-labeled indole ketole caswell no. 498B indole, 14C-labeled 14C-labeled indole indole (substance) indole, 14c-labeled epa pesticide chemical code 025000
Molecular Weight (g/mol):
117.15
Molecular Formula:
C8H7N
InChi Key:
SIKJAQJRHWYJAI-UHFFFAOYSA-N
SMILES:
N1C=CC2=CC=CC=C12
1
–
3
of
3
results
1
Indole, 99+%
CAS: 120-72-9 MDL Number: MFCD00005607 InChI Key: SIKJAQJRHWYJAI-UHFFFAOYSA-N Synonym: indole,indol,2,3-benzopyrrole,1-benzazole,ketole,1-azaindene,benzopyrrole,2,3-benzopyrole,caswell no. 498b,indol german PubChem CID: 798 ChEBI: CHEBI:16881 IUPAC Name: 1H-indole SMILES: C1=CC=C2C(=C1)C=CN2
2
Indole, 99%
CAS: 120-72-9 Molecular Formula: C8H7N Molecular Weight (g/mol): 117.151 MDL Number: MFCD00005607 InChI Key: SIKJAQJRHWYJAI-UHFFFAOYSA-N Synonym: indole,indol,2,3-benzopyrrole,1-benzazole,ketole,1-azaindene,benzopyrrole,2,3-benzopyrole,caswell no. 498b,indol german PubChem CID: 798 ChEBI: CHEBI:16881 IUPAC Name: 1H-indole SMILES: C1=CC=C2C(=C1)C=CN2