CAS RN 120162-55-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Azimsulfuron 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Azimsulfuron, TRC
CAS: 120162-55-2 Molecular Formula: C13 H16 N10 O5 S Molecular Weight (g/mol): 424.4 Synonym: N-[[(4,6-Dimethoxy-2-pyrimidinyl)amino]carbonyl]-1-methyl-4-(2-methyl-2H-tetrazol-5-yl)-1H-pyrazole-5-sulfonamide,Azimsulfuron,DPX 47,DPX-A 8947,Gulliver,IN-A 8947 IUPAC Name: 1-(4,6-dimethoxypyrimidin-2-yl)-3-[2-methyl-4-(2-methyltetrazol-5-yl)pyrazol-3-yl]sulfonylurea SMILES: COc1cc(OC)nc(NC(=O)NS(=O)(=O)c2c(cnn2C)c3nnn(C)n3)n1